missing translation for 'onlineSavingsMsg'
Learn More

Tetrapropylammonium Hydroxide, 10% in Water

Catalog No. AC420721000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420721000 100 g Plastic bottle
AC420725000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC420721000 Supplier Thermo Scientific Chemicals Supplier No. 420721000

CAS: 4499-86-9 | C12H29NO | 203.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 4499-86-9 , 7732-18-5
Molecular Formula C12H29NO
Molecular Weight (g/mol) 203.37
MDL Number MFCD00009360
InChI Key LPSKDVINWQNWFE-UHFFFAOYSA-M
Synonym tetrapropylammonium hydroxide, tetra-n-propylammonium hydroxide, 1-propanaminium, n,n,n-tripropyl-, hydroxide, tpaoh, tetrapropylammonium oxide, unii-t686luy7nk, tetrapropylazanium hydroxide, t686luy7nk, ammonium, tetrapropyl-, hydroxide, n,n,n-tripropyl-1-propanaminium hydroxide
PubChem CID 20586
IUPAC Name tetrapropylazanium hydroxide
SMILES [OH-].CCC[N+](CCC)(CCC)CCC

Specifications

Color Colorless
Boiling Point 100.0°C to 102.0°C
Linear Formula (CH3CH2CH2)4NOH
Density 1.0000g/mL
Solubility Information Solubility in water: soluble in water.
Formula Weight 203.37
Percent Purity 9 to 11%
Packaging Plastic bottle
Specific Gravity 1
Quantity 100 g
Physical Form Liquid
Chemical Name or Material Tetrapropylammonium hydroxide, 10% in Water
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-800-6
RTECSNumber BS8400000
TSCA TSCA

RUO – Research Use Only