missing translation for 'onlineSavingsMsg'
Learn More

Tetrapropylammonium Bromide 98.0+%, TCI America™

Supplier:  TCI America T0694500G

 View more versions of this product

Catalog No. T0694500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tetrapropylammonium Bromide
White
MFCD00011840
tetrapropylammonium bromide, tetra-n-propylammonium bromide, 1-propanaminium, n,n,n-tripropyl-, bromide, tetrapropylazanium bromide, tpabr, n,n,n-tripropyl-1-propanaminium bromide, n,n,n-tripropylpropan-1-aminium bromide, tetrapropylammonium bromide tpabr, 1-propanaminium, n,n,n-tripropyl-, bromide 1:1, tetra propyl ammonium bromide
CCC[N+](CCC)(CCC)CCC.[Br-]
266.267
CHEBI:55318
≥98.0% (T)
1941-30-6
C12H28BrN
500 g
BGQMOFGZRJUORO-UHFFFAOYSA-M
tetrapropylazanium;bromide
74745
266.27
Crystalline Powder

Chemical Identifiers

1941-30-6
266.267
BGQMOFGZRJUORO-UHFFFAOYSA-M
74745
tetrapropylazanium;bromide
C12H28BrN
MFCD00011840
tetrapropylammonium bromide, tetra-n-propylammonium bromide, 1-propanaminium, n,n,n-tripropyl-, bromide, tetrapropylazanium bromide, tpabr, n,n,n-tripropyl-1-propanaminium bromide, n,n,n-tripropylpropan-1-aminium bromide, tetrapropylammonium bromide tpabr, 1-propanaminium, n,n,n-tripropyl-, bromide 1:1, tetra propyl ammonium bromide
CHEBI:55318
CCC[N+](CCC)(CCC)CCC.[Br-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0186

RTECSNumber : BS8390000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions