missing translation for 'onlineSavingsMsg'
Learn More

Tetrapropylammonium Bromide 98.0+%, TCI America™

Catalog No. p-7136397
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T069425G 25 g
T0694500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T069425G Supplier TCI America Supplier No. T069425G
Only null left

Chemical Identifiers

CAS 1941-30-6
Molecular Formula C12H28BrN
Molecular Weight (g/mol) 266.267
MDL Number MFCD00011840
InChI Key BGQMOFGZRJUORO-UHFFFAOYSA-M
Synonym tetrapropylammonium bromide, tetra-n-propylammonium bromide, 1-propanaminium, n,n,n-tripropyl-, bromide, tetrapropylazanium bromide, tpabr, n,n,n-tripropyl-1-propanaminium bromide, n,n,n-tripropylpropan-1-aminium bromide, tetrapropylammonium bromide tpabr, 1-propanaminium, n,n,n-tripropyl-, bromide 1:1, tetra propyl ammonium bromide
PubChem CID 74745
ChEBI CHEBI:55318
IUPAC Name tetrapropylazanium;bromide
SMILES CCC[N+](CCC)(CCC)CCC.[Br-]

Specifications

Color White
Quantity 25 g
Formula Weight 266.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrapropylammonium Bromide
EINECSNumber (2)-0186
RTECSNumber BS8390000
TSCA Yes