accessibility menu, dialog, popup

UserName

CAS RN 1941-30-6

IUPAC Name: tetrapropylazanium bromide
Synonyms: tetrapropylammonium bromide
Molecular Weight (g/mol): 266.27
Molecular Formula: C12H28BrN
InChi Key: BGQMOFGZRJUORO-UHFFFAOYSA-M
SMILES: [Br-].CCC[N+](CCC)(CCC)CCC
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (7)
  • (4)
  • (5)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (4)