missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraphenylcyclopentadienone 98.0+%, TCI America™
Supplier: TCI America T10625G
Specifications
| Tetraphenylcyclopentadienone | |
| 217°C | |
| C29H20O | |
| 5 g | |
| PLGPSDNOLCVGSS-UHFFFAOYSA-N | |
| 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one | |
| 68068 | |
| ≥98.0% (HPLC) |
| 479-33-4 | |
| Black | |
| MFCD00001407 | |
| tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone | |
| C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 | |
| 384.478 | |
| 384.48 | |
| Crystalline Powder |
Chemical Identifiers
| 479-33-4 | |
| 384.478 | |
| PLGPSDNOLCVGSS-UHFFFAOYSA-N | |
| 68068 | |
| C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 |
| C29H20O | |
| MFCD00001407 | |
| tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone | |
| 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one |
Safety and Handling
TSCA : Yes