accessibility menu, dialog, popup

UserName

O

CAS RN 479-33-4

IUPAC Name: tetraphenylcyclopenta-2,4-dien-1-one
Synonyms: tetraphenylcyclopentadienone
Molecular Weight (g/mol): 384.48
Molecular Formula: C29H20O
InChi Key: PLGPSDNOLCVGSS-UHFFFAOYSA-N
SMILES: O=C1C(=C(C(=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)