missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraphenylcyclopentadienone 98.0+%, TCI America™
$89.48 - $257.24
Chemical Identifiers
| CAS | 479-33-4 |
|---|---|
| Molecular Formula | C29H20O |
| Molecular Weight (g/mol) | 384.478 |
| MDL Number | MFCD00001407 |
| InChI Key | PLGPSDNOLCVGSS-UHFFFAOYSA-N |
| Synonym | tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone |
| PubChem CID | 68068 |
| IUPAC Name | 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one |
| SMILES | C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 |
Chemical Identifiers
| 479-33-4 | |
| 384.478 | |
| PLGPSDNOLCVGSS-UHFFFAOYSA-N | |
| 68068 | |
| C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 |
| C29H20O | |
| MFCD00001407 | |
| tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone | |
| 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one |
Specifications
| 479-33-4 | |
| Black | |
| MFCD00001407 | |
| tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone | |
| C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 | |
| 384.478 | |
| 384.48 | |
| Crystalline Powder |
| 217°C | |
| C29H20O | |
| 5 g | |
| PLGPSDNOLCVGSS-UHFFFAOYSA-N | |
| 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one | |
| 68068 | |
| ≥98.0% (HPLC) | |
| Tetraphenylcyclopentadienone |
Safety and Handling
TSCA : Yes