Learn More
Tetraphenylcyclopentadienone, 98%
CAS: 479-33-4 | C29H20O | 384.478 g/mol
Supplier: Thermo Scientific Chemicals A1275622
Description
It is an easily made building block for many organic and organometallic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an easily made building block for many organic and organometallic compounds.
Solubility
Soluble in Benzene. Insoluble in water.
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications
| Tetraphenylcyclopentadienone | |
| 217°C to 219°C | |
| MFCD00001407 | |
| 683115 | |
| Soluble in Benzene. Insoluble in water. | |
| C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 | |
| 384.478 | |
| 384.48 |
| 479-33-4 | |
| C29H20O | |
| 100 g | |
| tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone | |
| PLGPSDNOLCVGSS-UHFFFAOYSA-N | |
| 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one | |
| 68068 | |
| 98% |
Chemical Identifiers
| 479-33-4 | |
| 384.478 | |
| PLGPSDNOLCVGSS-UHFFFAOYSA-N | |
| 68068 | |
| C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 |
| C29H20O | |
| MFCD00001407 | |
| tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone | |
| 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one |
Safety and Handling
EINECSNumber : 207-530-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only