missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylcyclopentadienone, 98%

Catalog No. p-7020708
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1275606 5 g
AAA1275614 25 g
AAA1275622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1275606 Supplier Thermo Scientific Chemicals Supplier No. A1275606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 479-33-4 | C29H20O | 384.478 g/mol

It is an easily made building block for many organic and organometallic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an easily made building block for many organic and organometallic compounds.

Solubility
Soluble in Benzene. Insoluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 479-33-4
Molecular Formula C29H20O
Molecular Weight (g/mol) 384.478
MDL Number MFCD00001407
InChI Key PLGPSDNOLCVGSS-UHFFFAOYSA-N
Synonym tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone
PubChem CID 68068
IUPAC Name 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one
SMILES C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5

Specifications

Melting Point 217°C to 219°C
Quantity 5 g
Beilstein 683115
Solubility Information Soluble in Benzene. Insoluble in water.
Formula Weight 384.48
Percent Purity 98%
Chemical Name or Material Tetraphenylcyclopentadienone
EINECSNumber 207-530-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only