missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylthiuram Monosulfide 98.0+%, TCI America™

Catalog No. p-7136113
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T015725G 25 g
T0157500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T015725G Supplier TCI America Supplier No. T015725G
Only null left

Chemical Identifiers

CAS 97-74-5
Molecular Formula C6H12N2S3
Molecular Weight (g/mol) 208.356
MDL Number MFCD00014870
InChI Key REQPQFUJGGOFQL-UHFFFAOYSA-N
Synonym tetramethylthiuram monosulfide, bis dimethylthiocarbamoyl sulfide, monothiuram, unads, mono-thiurad, tmtm, thiuram mm, aceto tmtm, cyuram ms, ekagom tm
PubChem CID 7347
IUPAC Name dimethylcarbamothioyl N,N-dimethylcarbamodithioate
SMILES CN(C)C(=S)SC(=S)N(C)C

Specifications

Melting Point 110°C
Color Yellow
Quantity 25 g
UN Number 3077
Formula Weight 208.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylthiuram Monosulfide
EINECSNumber (2)-1820
RTECSNumber WQ1750000
TSCA Yes