Learn More
Tetramethylthiuram monosulfide, 97%
CAS: 97-74-5 | C6H12N2S3 | 208.356 g/mol
$125.34 - $125.34
Chemical Identifiers
| CAS | 97-74-5 |
|---|---|
| Molecular Formula | C6H12N2S3 |
| Molecular Weight (g/mol) | 208.356 |
| MDL Number | MFCD00014870 |
| InChI Key | REQPQFUJGGOFQL-UHFFFAOYSA-N |
| Synonym | tetramethylthiuram monosulfide, bis dimethylthiocarbamoyl sulfide, monothiuram, unads, mono-thiurad, tmtm, thiuram mm, aceto tmtm, cyuram ms, ekagom tm |
| PubChem CID | 7347 |
| IUPAC Name | dimethylcarbamothioyl N,N-dimethylcarbamodithioate |
| SMILES | CN(C)C(=S)SC(=S)N(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1725922
|
Thermo Scientific Chemicals
A1725922 |
100 g |
Each for $125.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 97-74-5 | |
| 208.356 | |
| REQPQFUJGGOFQL-UHFFFAOYSA-N | |
| 7347 | |
| CN(C)C(=S)SC(=S)N(C)C |
| C6H12N2S3 | |
| MFCD00014870 | |
| tetramethylthiuram monosulfide, bis dimethylthiocarbamoyl sulfide, monothiuram, unads, mono-thiurad, tmtm, thiuram mm, aceto tmtm, cyuram ms, ekagom tm | |
| dimethylcarbamothioyl N,N-dimethylcarbamodithioate |
Specifications
| 97-74-5 | |
| 1.37 | |
| MFCD00014870 | |
| 1775650 | |
| REQPQFUJGGOFQL-UHFFFAOYSA-N | |
| dimethylcarbamothioyl N,N-dimethylcarbamodithioate | |
| 7347 | |
| 97% |
| 106°C to 110°C | |
| C6H12N2S3 | |
| 100 g | |
| tetramethylthiuram monosulfide, bis dimethylthiocarbamoyl sulfide, monothiuram, unads, mono-thiurad, tmtm, thiuram mm, aceto tmtm, cyuram ms, ekagom tm | |
| CN(C)C(=S)SC(=S)N(C)C | |
| 208.356 | |
| 208.37 | |
| Tetramethylthiuram monosulfide |
Safety and Handling
GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
H302-H317
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 202-605-7
RTECSNumber : WQ1750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only