missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylhydroquinone 95.0+%, TCI America™
$552.52 - $1749.98
Chemical Identifiers
| CAS | 527-18-4 |
|---|---|
| Molecular Formula | C10H14O2 |
| Molecular Weight (g/mol) | 166.22 |
| MDL Number | MFCD00045781 |
| InChI Key | SUNVJLYYDZCIIK-UHFFFAOYSA-N |
| Synonym | Dihydroxydurene, Durohydroquinone, 1,4-Dihydroxy-2,3,5,6-tetramethylbenzene, 2,3,5,6-Tetramethyl-1,4-benzenediol |
| PubChem CID | 136346 |
| IUPAC Name | 2,3,5,6-tetramethylbenzene-1,4-diol |
| SMILES | CC1=C(C(=C(C(=C1O)C)C)O)C |
Chemical Identifiers
| 527-18-4 | |
| 166.22 | |
| SUNVJLYYDZCIIK-UHFFFAOYSA-N | |
| 136346 | |
| CC1=C(C(=C(C(=C1O)C)C)O)C |
| C10H14O2 | |
| MFCD00045781 | |
| Dihydroxydurene, Durohydroquinone, 1,4-Dihydroxy-2,3,5,6-tetramethylbenzene, 2,3,5,6-Tetramethyl-1,4-benzenediol | |
| 2,3,5,6-tetramethylbenzene-1,4-diol |
Specifications
| 527-18-4 | |
| C10H14O2 | |
| 1 g | |
| SUNVJLYYDZCIIK-UHFFFAOYSA-N | |
| 2,3,5,6-tetramethylbenzene-1,4-diol | |
| 136346 | |
| ≥95.0% (GC) | |
| Tetramethylhydroquinone |
| 235°C | |
| MFCD00045781 | |
| Dihydroxydurene, Durohydroquinone, 1,4-Dihydroxy-2,3,5,6-tetramethylbenzene, 2,3,5,6-Tetramethyl-1,4-benzenediol | |
| CC1=C(C(=C(C(=C1O)C)C)O)C | |
| 166.22 | |
| 166.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No