accessibility menu, dialog, popup

UserName

CH3CH3OHCH3H3CHO

CAS RN 527-18-4

IUPAC Name: tetramethylbenzene-1,4-diol
Synonyms: tetramethylbenzene-1,4-diol
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: SUNVJLYYDZCIIK-UHFFFAOYSA-N
SMILES: CC1=C(C)C(O)=C(C)C(C)=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results