missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium p-Toluenesulfonate 99.0+%, TCI America™

Catalog No. T084325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T084325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T084325G Supplier TCI America Supplier No. T084325G

Chemical Identifiers

CAS 3983-91-3
Molecular Formula C11H19NO3S
Molecular Weight (g/mol) 245.34
MDL Number MFCD00043173
InChI Key FHVCZJGBXWNGIZ-UHFFFAOYSA-M
PubChem CID 6451622
IUPAC Name tetramethylazanium 4-methylbenzene-1-sulfonate
SMILES C[N+](C)(C)C.CC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 245.34
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylammonium p-Toluenesulfonate
EINECSNumber (2)-0186
TSCA No