accessibility menu, dialog, popup

UserName

CH3SOOOCH3NCH3CH3H3C

CAS RN 3983-91-3

IUPAC Name: tetramethylazanium 4-methylbenzene-1-sulfonate
Synonyms: tetramethylammonium ion tosylate
Molecular Weight (g/mol): 245.34
Molecular Formula: C11H19NO3S
InChi Key: FHVCZJGBXWNGIZ-UHFFFAOYSA-M
SMILES: C[N+](C)(C)C.CC1=CC=C(C=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results