missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Hydroxide, 10% (w/w) Aqueous, Ricca Chemical

Catalog No. 838616
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
838616 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 838616 Supplier Ricca Chemical Company Supplier No. 838616

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-59-2 , 7732-18-5
Molecular Formula C4H13NO
Molecular Weight (g/mol) 91.154
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
Synonym tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
PubChem CID 60966
IUPAC Name tetramethylazanium;hydroxide
SMILES C[N+](C)(C)C.[OH-]

Specifications

Color Colorless
CAS Min % 58.0, 39.67
CAS Max % 61.0, 41.29
Percent Purity 10%
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
pH >13
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 10% (w/w)
Chemical Name or Material Tetramethylammonium Hydroxide
Show More Show Less