missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium Chloride, High Purity, 99%, Spectrum™ Chemical
$285.53 - $19534.44
Chemical Identifiers
| CAS | 75-57-0 |
|---|---|
| Molecular Formula | C4H12ClN |
| Molecular Weight (g/mol) | 109.60 |
| InChI Key | OKIZCWYLBDKLSU-UHFFFAOYSA-M |
| IUPAC Name | tetramethylazanium chloride |
| SMILES | [Cl-].C[N+](C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18614371
|
Spectrum Chemical Mfg Cor
T1036100GM |
100 g | Amber Glass Bottle |
Each for $285.53
|
|
||||
|
18614372
|
Spectrum Chemical Mfg Cor
T1036500GM |
500 g | Amber Glass Bottle |
Each for $571.62
|
|
||||
|
18614373
|
Spectrum Chemical Mfg Cor
T103650KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.