accessibility menu, dialog, popup

UserName

CAS RN 75-57-0

IUPAC Name: tetramethylazanium chloride
Synonyms: tetramethylammonium chloride
Molecular Weight (g/mol): 109.6
Molecular Formula: C4H12ClN
InChi Key: OKIZCWYLBDKLSU-UHFFFAOYSA-M
SMILES: [Cl-].C[N+](C)(C)C
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (5)
  • (3)
  • (5)
Physical Form 
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
Grade 
  • (3)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (7)
  • (2)
  • (7)
  • (2)
  • (6)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (3)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (4)