missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Chloride, High Purity, 99%, Spectrum™ Chemical

Catalog No. 18614371
Change view
Click to view available options
Quantity:
100 g
500 g
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614371 100 g Amber Glass Bottle
18614372 500 g Amber Glass Bottle
18614373 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18614371 Supplier Spectrum Chemical Mfg Cor Supplier No. T1036100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 75-57-0
Molecular Formula C4H12ClN
Molecular Weight (g/mol) 109.60
InChI Key OKIZCWYLBDKLSU-UHFFFAOYSA-M
IUPAC Name tetramethylazanium chloride
SMILES [Cl-].C[N+](C)(C)C

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 0.99
Grade High Purity

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.