missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium chloride, For ion pair chromatography (AT), ≥99.0% (AT), MilliporeSigma™ Supelco™

$155.05 - $622.10

Chemical Identifiers

MDL Number MFCD00011628
Synonym TMA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111019736
View Documents
Merck Emd Millipore
7420250GF
50 g
Each for $155.05
Only null left
 
111018327
View Documents
Merck Emd Millipore
74202250GF
250 g
Each for $622.10
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00011628
TMA

Specifications

>300°C
50 g
MFCD00011628
109.60
Ion Pair Chromatography
75-57-0
(CH3)4 N(Cl)
TMA
≥99.0% (AT)
Videos
Safety and Handling

Safety and Handling

RTECSNumber : BS7700000

SDS
missing translation for 'documents'

missing translation for 'documents'