missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium chloride, For ion pair chromatography (AT), ≥99.0% (AT), MilliporeSigma™ Supelco™

Catalog No. 111019736
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019736 50 g
111018327 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019736 Supplier Merck Emd Millipore Supplier No. 7420250GF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00011628
Synonym TMA

Specifications

Melting Point >300°C
CAS 75-57-0
Physical Form Crystals
Quantity 50 g
Linear Formula (CH3)4 N(Cl)
Molecular Weight (g/mol) 109.60
Formula Weight 109.60
Percent Purity ≥99.0% (AT)
Grade Ion Pair Chromatography
Chemical Name or Material Tetramethylammonium Chloride
RTECSNumber BS7700000