missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Chloride, Crystal, 98.0 wt.%, Spectrum™ Chemical

$231.07 - $12231.18

Chemical Identifiers

CAS 75-57-0
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614367
View Documents
Spectrum Chemical Mfg Cor
T1068100GM
100 g Amber Glass Bottle
Each for $231.07
Only null left
 
18614368
View Documents
Spectrum Chemical Mfg Cor
T1068500GM
500 g Amber Glass Bottle
Each for $534.68
Only null left
 
18614369
View Documents
Spectrum Chemical Mfg Cor
T106825KG
2.5 kg Poly Pail
Each for $1,498.57
Only null left
 
18614370
View Documents
Spectrum Chemical Mfg Cor
T106850KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

75-57-0

Specifications

75-57-0
Amber Glass Bottle
98.0 wt.%
1
100 g
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.