missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Chloride, Crystal, 98.0 wt.%, Spectrum™ Chemical

Catalog No. 18614367
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614367 100 g Amber Glass Bottle
18614368 500 g Amber Glass Bottle
18614369 2.5 kg Poly Pail
18614370 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614367 Supplier Spectrum Chemical Mfg Cor Supplier No. T1068100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 75-57-0

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98.0 wt.%
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.