missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(4-bromophenyl)methane 95.0+%, TCI America™

Catalog No. p-7137499
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29601G 1 g
T29605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29601G Supplier TCI America Supplier No. T29601G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105309-59-9
Molecular Formula C25H16Br4
Molecular Weight (g/mol) 636.019
MDL Number MFCD19688472
InChI Key YBGIIZGNEOJSRF-UHFFFAOYSA-N
PubChem CID 11250692
IUPAC Name 1-bromo-4-[tris(4-bromophenyl)methyl]benzene
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)Br)(C3=CC=C(C=C3)Br)C4=CC=C(C=C4)Br)Br

Specifications

Melting Point 300°C
Color Yellow
Quantity 1 g
Formula Weight 636.02
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrakis(4-bromophenyl)methane