accessibility menu, dialog, popup

UserName

BrBrBrBr

CAS RN 105309-59-9

IUPAC Name: 1-bromo-4-[tris(4-bromophenyl)methyl]benzene
Synonyms: 1-bromo-4-[tris(4-bromophenyl)methyl]benzene
Molecular Weight (g/mol): 636.02
Molecular Formula: C25H16Br4
InChi Key: YBGIIZGNEOJSRF-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C(C1=CC=C(Br)C=C1)(C1=CC=C(Br)C=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results