missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, Anhydrous, septum-sealed, for applications requiring low water content, contains no preservative, >99.9%, Honeywell Burdick & Jackson™
Anhydrous, septum-sealed, for applications requiring low water content, contains no preservative, >99.9%
$2792.65 - $2792.65
Chemical Identifiers
| CAS | 109-99-9 |
|---|---|
| Molecular Formula | C4H8O |
| Molecular Weight (g/mol) | 72.11 |
| MDL Number | MFCD00005356 |
| InChI Key | WYURNTSHIVDZCO-UHFFFAOYSA-N |
| Synonym | tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano |
| PubChem CID | 8028 |
| ChEBI | CHEBI:26911 |
| IUPAC Name | oxolane |
| SMILES | C1CCOC1 |
Description
- Water content ranging from <10 ppm to <50 ppm
- Low residue
- Convenient and effective packaging ensures product quality
- Certificate of Analysis printed on the label
Chemical Identifiers
| 109-99-9 | |
| 72.11 | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| 8028 | |
| oxolane |
| C4H8O | |
| MFCD00005356 | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| CHEBI:26911 | |
| C1CCOC1 |
Specifications
| 109-99-9 | |
| 0.8892 g/cm3 | |
| 6 x 1 L | |
| Septum-sealed Bottle | |
| C4H8O | |
| UN2056 | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| C1CCOC1 | |
| 72.11 | |
| CHEBI:26911 | |
| >99.9% |
| 100.0000% | |
| 66°C | |
| −14°C | |
| anhydrous | |
| MFCD00005356 | |
| 102391 | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| oxolane | |
| 8028 | |
| 72.11 | |
| Tetrahydrofuran, Anhydrous, septum-sealed, for applications requiring low water content, contains no preservative, >99.9% |
Safety and Handling
ShelfLife : 365 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Tetrahydrofuran