missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Anhydrous, septum-sealed, for applications requiring low water content, contains no preservative, >99.9%, Honeywell Burdick & Jackson™

Catalog No. 6001668
Change view
Click to view available options
Quantity:
6 x 1 L
Packaging:
Septum-sealed Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001668 6 x 1 L Septum-sealed Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001668 Supplier Honeywell Chemicals Supplier No. AS3401
Only null left

Anhydrous, septum-sealed, for applications requiring low water content, contains no preservative, >99.9%

  • Water content ranging from <10 ppm to <50 ppm
  • Low residue
  • Convenient and effective packaging ensures product quality
  • Certificate of Analysis printed on the label

 

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

CAS Max % 100.0000%
Density 0.8892 g/cm3
Quantity 6 x 1 L
Flash Point −14°C
Packaging Septum-sealed Bottle
Physical Form anhydrous
UN Number UN2056
Beilstein 102391
Boiling Point 66°C
Formula Weight 72.11
Percent Purity >99.9%
Chemical Name or Material Tetrahydrofuran, Anhydrous, septum-sealed, for applications requiring low water content, contains no preservative, >99.9%
Show More Show Less
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Tetrahydrofuran