Learn More
Tetrahydrofuran, anhydrous, 99.8+%, unstab.
For Synthesis and Liquid Chromatography | CAS: 109-99-9 | C4H8O | 72.11 g/mol
Supplier: Thermo Scientific Chemicals 042254K2
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tetrahydrofuran | |
| 109-99-9 | |
| Colorless liquid | |
| 66°C | |
| -17°C (1°F) | |
| Liquid | |
| >99.8% | |
| MFCD00005356 | |
| 102391 | |
| Miscible with water. | |
| C1CCOC1 | |
| 72.11 | |
| CHEBI:26911 |
| °C to 108°C | |
| Colorless | |
| 0.889 | |
| 1 L | |
| Ether-like | |
| 1.407 | |
| C4H8O | |
| 200 hPa (150mm Hg) at 20°C | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| oxolane | |
| 8028 | |
| 72.11 |
Chemical Identifiers
| 109-99-9 | |
| 72.11 | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| 8028 | |
| oxolane |
| C4H8O | |
| MFCD00005356 | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| CHEBI:26911 | |
| C1CCOC1 |
Safety and Handling
Keep away from food, drink and animal feeding stuffs. Keep away from sources of ignition - No Smoking. Do not empty into drains. Take precautionary measures against static discharges. Wear suitable protective clothing and gloves. If swallowed seek medical advice immediately and show this container or label.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
H225-H302-H319-H335-H336-H351
DOTInformation : DOT Class: 3, Packaging Group: II
EINECSNumber : 203-726-8
RTECSNumber : LU5950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only