missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, anhydrous, 99.8+%, unstab.

Catalog No. p-4782180
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42254AK 250 mL
AA42254K2 1 L
AA42254K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42254AK Supplier Thermo Scientific Chemicals Supplier No. 42254AK
Only null left

For Synthesis and Liquid Chromatography | CAS: 109-99-9 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point °C to 108°C
Color Colorless
Appearance Colorless liquid
Density 0.889
Quantity 250 mL
Flash Point -17°C (1°F)
Odor Ether-like
Physical Form Liquid
Refractive Index 1.407
Assay Percent Range >99.8%
Vapor Pressure 200 hPa (150mm Hg) at 20°C
Beilstein 102391
Boiling Point 66°C
Solubility Information Miscible with water.
Formula Weight 72.11
Chemical Name or Material Tetrahydrofuran
Show More Show Less
Hazard Category H225-H302-H319-H335-H336-H351
Hazard Statement Keep away from food, drink and animal feeding stuffs. Keep away from sources of ignition - No Smoking. Do not empty into drains. Take precautionary measures against static discharges. Wear suitable protective clothing and gloves. If swallowed seek medical advice immediately and show this container or label.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 203-726-8
RTECSNumber LU5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only