Learn More
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Supplier: Thermo Scientific Chemicals 326970010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tetrahydrofuran | |
| Stabilized, water < 50ppm | |
| Colorless | |
| 66.0°C | |
| −21°C | |
| Liquid | |
| 1.407 | |
| 99.85% | |
| C4H8O | |
| 0.55 mPa.s (20°C) | |
| 17, 10 | |
| 15, 9356 | |
| Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone | |
| C1CCOC1 | |
| 72.11 | |
| CHEBI:26911 | |
| 99.85% |
| -108.4°C | |
| 109-99-9,128-37-0 | |
| 0.8800g/mL | |
| 1 L | |
| Glass Bottle | |
| Water <50ppm | |
| 0.001 % max. | |
| 0.88 | |
| MFCD00005356 | |
| 0.005% Max. (Coulometric) | |
| 01,1140; 02,398; 03,278; 05,649; 06,570 | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| oxolane | |
| 8028 | |
| 72.11 | |
| Extra Dry |
Chemical Identifiers
| 109-99-9 | |
| 72.11 | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| 8028 | |
| oxolane |
| C4H8O | |
| MFCD00005356 | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| CHEBI:26911 | |
| C1CCOC1 |
Safety and Handling
GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. Suspected of causing cancer.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Keep container tightly closed.
Danger
EINECSNumber : 203-726-8
RTECSNumber : LU5950000
TSCA : TSCA
RUO – Research Use Only