missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -108.4°C |
| Color | Colorless |
| Density | 0.8800g/mL |
| Boiling Point | 66.0°C |
| Quantity | 100 mL |
| Flash Point | −21°C |
| Packaging | AcroSeal™ Glass Bottle |
| Physical Form | Liquid |
| Refractive Index | 1.407 |
| Residue after Evaporation | 0.001 % max. |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. Suspected of causing cancer. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Keep container tightly closed. |
| EINECSNumber | 203-726-8 |
| RTECSNumber | LU5950000 |
| TSCA | TSCA |
RUO – Research Use Only