missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluoroterephthalonitrile 98.0+%, TCI America™

Catalog No. p-7136574
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10501G 1 g
T10505G 5 g
T105025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T10501G Supplier TCI America Supplier No. T10501G

Chemical Identifiers

CAS 1835-49-0
Molecular Formula C8F4N2
Molecular Weight (g/mol) 200.096
MDL Number MFCD00001776
InChI Key PCRSJGWFEMHHEW-UHFFFAOYSA-N
Synonym tetrafluoroterephthalonitrile, 2,3,5,6-tetrafluoroterephthalonitrile, diamond 2031, 2,3,5,6-tetrafluoro-1,4-benzenedicarbonitrile, 1,4-benzenedicarbonitrile, 2,3,5,6-tetrafluoro, 1,4-dicyanotetrafluorobenzene, terephthalonitrile, tetrafluoro, perfluoroterephthalonitrile, tetrafluoroterephthalodinitrile, tetrafluorobenzene-1,4-dicarbonitrile
PubChem CID 15783
IUPAC Name 2,3,5,6-tetrafluorobenzene-1,4-dicarbonitrile
SMILES C(#N)C1=C(C(=C(C(=C1F)F)C#N)F)F

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 200.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrafluoroterephthalonitrile
RTECSNumber CZ1955000
TSCA No