missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluorotetracyanoquinodimethane (purified by sublimation) 98.0+%, TCI America™

Catalog No. T1131100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1131100MG 100 mg
T11311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1131100MG Supplier TCI America Supplier No. T1131100MG

Chemical Identifiers

CAS 29261-33-4
Molecular Formula C12F4N4
Molecular Weight (g/mol) 276.154
MDL Number MFCD00042382
InChI Key IXHWGNYCZPISET-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 2,2'-perfluorocyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tcnqf4, 2,3,5,6-tetrafluorotetracyanoquinodimethane, f4-tcnq, tetrafluorotetracyanoquinodimethane, 2-4-dicyanomethylidene-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene propanedinitrile, f4tcnq, tetrafluorotetracyanoquinodimethane anion radical, tetrafluorotetracyanoquinodimethane purifiedbysubl
PubChem CID 2733307
IUPAC Name 2-[4-(dicyanomethylidene)-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
SMILES C(#N)C(=C1C(=C(C(=C(C#N)C#N)C(=C1F)F)F)F)C#N

Specifications

Melting Point 300°C
Quantity 100 mg
UN Number 3439
Formula Weight 276.15
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tetrafluorotetracyanoquinodimethane (purified by sublimation) [Organic Electronic Material]