accessibility menu, dialog, popup

UserName

FFFNNFNN

CAS RN 29261-33-4

IUPAC Name: 2-[4-(dicyanomethylidene)-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
Synonyms: 2-[4-(dicyanomethylidene)-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
Molecular Weight (g/mol): 276.15
Molecular Formula: C12F4N4
InChi Key: IXHWGNYCZPISET-UHFFFAOYSA-N
SMILES: FC1=C(F)C(=C(C#N)C#N)C(F)=C(F)C1=C(C#N)C#N
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)