missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluoroterephthalic Acid 97.0+%, TCI America™

Catalog No. T09305G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09305G 5 g
T09301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09305G Supplier TCI America Supplier No. T09305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 652-36-8
Molecular Formula C8H2F4O4
Molecular Weight (g/mol) 238.09
MDL Number MFCD00012301
InChI Key WFNRNCNCXRGUKN-UHFFFAOYSA-N
PubChem CID 69549
IUPAC Name tetrafluorobenzene-1,4-dicarboxylic acid
SMILES OC(=O)C1=C(F)C(F)=C(C(O)=O)C(F)=C1F

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 238.09
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrafluoroterephthalic Acid