accessibility menu, dialog, popup

UserName

HOOFFOHOFF

CAS RN 652-36-8

IUPAC Name: tetrafluorobenzene-1,4-dicarboxylic acid
Synonyms: tetrafluoroterephthalic acid
Molecular Weight (g/mol): 238.09
Molecular Formula: C8H2F4O4
InChi Key: WFNRNCNCXRGUKN-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(F)=C(C(O)=O)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results