missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluorosuccinic Acid 98.0+%, TCI America™

Supplier:  TCI America T162125G

 View more versions of this product

Catalog No. T162125G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tetrafluorosuccinic Acid
116°C
120°C
MFCD00042430
tetrafluorosuccinic acid, perfluorosuccinic acid, butanedioic acid, tetrafluoro, succinic acid, perfluoro, succinic acid, tetrafluoro, tetrafluorobutanedioic acid, butanedioic acid, 2,2,3,3-tetrafluoro, 2,2,3,3-tetrafluorosuccinic acid, butanedioic acid,2,2,3,3-tetrafluoro, perfluorosuccinate
C(=O)(C(C(C(=O)O)(F)F)(F)F)O
190.05
190.05
Crystalline Powder
377-38-8
White-Yellow
C4H2F4O4
25 g
YUDUFRYTKFGQCL-UHFFFAOYSA-N
2,2,3,3-tetrafluorobutanedioic acid
67833
≥98.0% (T)

Chemical Identifiers

377-38-8
190.05
YUDUFRYTKFGQCL-UHFFFAOYSA-N
2,2,3,3-tetrafluorobutanedioic acid
C4H2F4O4
MFCD00042430
67833
C(=O)(C(C(C(=O)O)(F)F)(F)F)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : WN0725000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions