accessibility menu, dialog, popup

UserName

CAS RN 377-38-8

IUPAC Name: tetrafluorobutanedioic acid
Synonyms: butanedioic acid, tetrafluoro-
Molecular Weight (g/mol): 190.05
Molecular Formula: C4H2F4O4
InChi Key: YUDUFRYTKFGQCL-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(O)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)