missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrafluorosuccinic Acid 98.0+%, TCI America™
$121.16 - $467.87
Chemical Identifiers
| CAS | 377-38-8 |
|---|---|
| Molecular Formula | C4H2F4O4 |
| Molecular Weight (g/mol) | 190.05 |
| MDL Number | MFCD00042430 |
| InChI Key | YUDUFRYTKFGQCL-UHFFFAOYSA-N |
| PubChem CID | 67833 |
| IUPAC Name | 2,2,3,3-tetrafluorobutanedioic acid |
| SMILES | C(=O)(C(C(C(=O)O)(F)F)(F)F)O |
Chemical Identifiers
| 377-38-8 | |
| 190.05 | |
| YUDUFRYTKFGQCL-UHFFFAOYSA-N | |
| 2,2,3,3-tetrafluorobutanedioic acid |
| C4H2F4O4 | |
| MFCD00042430 | |
| 67833 | |
| C(=O)(C(C(C(=O)O)(F)F)(F)F)O |
Specifications
| 377-38-8 | |
| White-Yellow | |
| C4H2F4O4 | |
| 5 g | |
| YUDUFRYTKFGQCL-UHFFFAOYSA-N | |
| 2,2,3,3-tetrafluorobutanedioic acid | |
| 67833 | |
| ≥98.0% (T) | |
| Tetrafluorosuccinic Acid |
| 116°C | |
| 120°C | |
| MFCD00042430 | |
| tetrafluorosuccinic acid, perfluorosuccinic acid, butanedioic acid, tetrafluoro, succinic acid, perfluoro, succinic acid, tetrafluoro, tetrafluorobutanedioic acid, butanedioic acid, 2,2,3,3-tetrafluoro, 2,2,3,3-tetrafluorosuccinic acid, butanedioic acid,2,2,3,3-tetrafluoro, perfluorosuccinate | |
| C(=O)(C(C(C(=O)O)(F)F)(F)F)O | |
| 190.05 | |
| 190.05 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : WN0725000
TSCA : Yes