missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylsilane, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 313701000
| Quantity | 100g |
|---|
Chemical Identifiers
| 631-36-7 | |
| 144.34 | |
| silane, tetraethyl, tetraethylsilicon, tetraethylsilane, acmc-209ndb, c2h5 4si | |
| tetraethylsilane |
| C8H20Si | |
| VCZQFJFZMMALHB-UHFFFAOYSA-N | |
| 12426 | |
| CC[Si](CC)(CC)CC |
Specifications
| Tetraethylsilane | |
| 0.7600g/mL | |
| 26°C | |
| 96% min. (GC) | |
| 1.4250 to 1.4280 | |
| 100g | |
| silane, tetraethyl, tetraethylsilicon, tetraethylsilane, acmc-209ndb, c2h5 4si | |
| VCZQFJFZMMALHB-UHFFFAOYSA-N | |
| tetraethylsilane | |
| 12426 | |
| 97% |
| 631-36-7 | |
| 153.0°C to 154.0°C | |
| Authentic | |
| C8H20Si | |
| Si(C2H5)4 | |
| 0.76 | |
| Solubility in water: insoluble. | |
| CC[Si](CC)(CC)CC | |
| 144.34 | |
| 144.34 |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
GHS Signal Word: Warning
EINECSNumber : 211-155-