accessibility menu, dialog, popup

UserName

CH3SiH3CH3CH3C

CAS RN 631-36-7

IUPAC Name: tetraethylsilane
Synonyms: tetraethylsilane
Molecular Weight (g/mol): 144.33
Molecular Formula: C8H20Si
InChi Key: VCZQFJFZMMALHB-UHFFFAOYSA-N
SMILES: CC[Si](CC)(CC)CC
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)