missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylsilane, 97%, Thermo Scientific™

Catalog No. p-7077230
Change view
Click to view available options
Quantity:
5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC313700050 5g
AC313700250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC313700050 Supplier Thermo Scientific Chemicals Supplier No. 313700050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 631-36-7
Molecular Formula C8H20Si
Molecular Weight (g/mol) 144.34
InChI Key VCZQFJFZMMALHB-UHFFFAOYSA-N
Synonym silane, tetraethyl, tetraethylsilicon, tetraethylsilane, acmc-209ndb, c2h5 4si
PubChem CID 12426
IUPAC Name tetraethylsilane
SMILES CC[Si](CC)(CC)CC

Specifications

Density 0.7600g/mL
Boiling Point 153.0°C to 154.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4250 to 1.4280
Linear Formula Si(C2H5)4
Quantity 5g
Specific Gravity 0.76
Solubility Information Solubility in water: insoluble.
Formula Weight 144.34
Percent Purity 97%
Chemical Name or Material Tetraethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
EINECSNumber 211-155-

RUO – Research Use Only