missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylene glycol monooctyl ether

Reagent for solubilizing membrane proteins | CAS: 19327-39-0 | C16H34O5 | 306.443 g/mol

Supplier:  Thermo Scientific Chemicals J6249606

 View more versions of this product

Catalog No. AAJ6249606

Only null left
Explore available promotions

Description

Description

Reagent for solubilizing membrane proteins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetraethylene glycol monooctyl ether
Colorless
C16H34O5
MFCD00043033
1781226
FEOZZFHAVXYAMB-UHFFFAOYSA-N
2-[2-[2-(2-octoxyethoxy)ethoxy]ethoxy]ethanol
5414
306.44
19327-39-0
0.969
CH3(CH2)7(OCH2CH2)4OH
5 g
3,6,9,12-tetraoxaeicosan-1-ol, 3,6,9,12-tetraoxaicosan-1-ol, tetraethylene glycol monooctyl ether, hydroxyethyloxy tri ethyloxy octane, tetraethylene glycol monoctyl ether, c8e, octyltetraglycol, octyltetraethyleneoxide, d06pvf, octyl tetraethylene glycol ether
CCCCCCCCOCCOCCOCCOCCO
306.443
CHEBI:41323
Liquid

Chemical Identifiers

19327-39-0
306.443
FEOZZFHAVXYAMB-UHFFFAOYSA-N
5414
2-[2-[2-(2-octoxyethoxy)ethoxy]ethoxy]ethanol
C16H34O5
MFCD00043033
3,6,9,12-tetraoxaeicosan-1-ol, 3,6,9,12-tetraoxaicosan-1-ol, tetraethylene glycol monooctyl ether, hydroxyethyloxy tri ethyloxy octane, tetraethylene glycol monoctyl ether, c8e, octyltetraglycol, octyltetraethyleneoxide, d06pvf, octyl tetraethylene glycol ether
CHEBI:41323
CCCCCCCCOCCOCCOCCOCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only