accessibility menu, dialog, popup

UserName

CAS RN 19327-39-0

IUPAC Name: 3,6,9,12-tetraoxaicosan-1-ol
Synonyms: 3,6,9,12-tetraoxaeicosan-1-ol (hydroxyethyloxy)tri(ethyloxy)octane
Molecular Weight (g/mol): 306.44
Molecular Formula: C16H34O5
InChi Key: FEOZZFHAVXYAMB-UHFFFAOYSA-N
SMILES: CCCCCCCCOCCOCCOCCOCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results