missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylene glycol monooctyl ether

Catalog No. p-7042431
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6249603 1 g
AAJ6249606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6249603 Supplier Thermo Scientific Chemicals Supplier No. J6249603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent for solubilizing membrane proteins | CAS: 19327-39-0 | C16H34O5 | 306.443 g/mol

Reagent for solubilizing membrane proteins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19327-39-0
Molecular Formula C16H34O5
Molecular Weight (g/mol) 306.443
MDL Number MFCD00043033
InChI Key FEOZZFHAVXYAMB-UHFFFAOYSA-N
Synonym 3,6,9,12-tetraoxaeicosan-1-ol, 3,6,9,12-tetraoxaicosan-1-ol, tetraethylene glycol monooctyl ether, hydroxyethyloxy tri ethyloxy octane, tetraethylene glycol monoctyl ether, c8e, octyltetraglycol, octyltetraethyleneoxide, d06pvf, octyl tetraethylene glycol ether
PubChem CID 5414
ChEBI CHEBI:41323
IUPAC Name 2-[2-[2-(2-octoxyethoxy)ethoxy]ethoxy]ethanol
SMILES CCCCCCCCOCCOCCOCCOCCO

Specifications

Color Colorless
Density 0.969
Linear Formula CH3(CH2)7(OCH2CH2)4OH
Quantity 1 g
Beilstein 1781226
Formula Weight 306.44
Physical Form Liquid
Chemical Name or Material Tetraethylene glycol monooctyl ether
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only