missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. p-7136507
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T093725G 25 g
T0937500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T093725G Supplier TCI America Supplier No. T093725G
Only null left

Chemical Identifiers

CAS 733-44-8
Molecular Formula C15H27NO3S
Molecular Weight (g/mol) 301.445
MDL Number MFCD00011831
InChI Key QKFFSWPNFCXGIQ-UHFFFAOYSA-M
Synonym tetraethylammonium p-toluenesulfonate, tetraethylammonium tosylate, ethanaminium, n,n,n-triethyl-, salt with 4-methylbenzenesulfonic acid 1:1, tetraethylammonium 4-methylbenzenesulfonate, ethanaminium, n,n,n-triethyl-, 4-methylbenzenesulfonate 1:1, 4-methylbenzenesulfonate; tetraethylazanium, et4nots, tetraethylammonium p-toluenesulphonate, acmc-209oql, n,n,n-triethylethanaminium-4-methylbenzolsulfonat
PubChem CID 101846
IUPAC Name 4-methylbenzenesulfonate;tetraethylazanium
SMILES CC[N+](CC)(CC)CC.CC1=CC=C(C=C1)S(=O)(=O)[O-]

Specifications

Melting Point 114°C
Color White
Quantity 25 g
Formula Weight 301.45
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylammonium p-Toluenesulfonate
EINECSNumber (2)-0186
TSCA Yes