CAS RN 733-44-8
CAS RN 733-44-8
IUPAC Name:
tetraethylazanium 4-methylbenzene-1-sulfonate
Synonyms:
tetraethylammonium tosylate
Molecular Weight (g/mol):
301.45
Molecular Formula:
C15H27NO3S
InChi Key:
QKFFSWPNFCXGIQ-UHFFFAOYSA-M
SMILES:
CC[N+](CC)(CC)CC.CC1=CC=C(C=C1)S([O-])(=O)=O
1
–
1
of
1
results
1
Tetraethylammonium p-Toluenesulfonate 98.0+%, TCI America™
CAS: 733-44-8 Molecular Formula: C15H27NO3S Molecular Weight (g/mol): 301.445 MDL Number: MFCD00011831 InChI Key: QKFFSWPNFCXGIQ-UHFFFAOYSA-M Synonym: tetraethylammonium p-toluenesulfonate,tetraethylammonium tosylate,ethanaminium, n,n,n-triethyl-, salt with 4-methylbenzenesulfonic acid 1:1,tetraethylammonium 4-methylbenzenesulfonate,ethanaminium, n,n,n-triethyl-, 4-methylbenzenesulfonate 1:1,4-methylbenzenesulfonate; tetraethylazanium,et4nots,tetraethylammonium p-toluenesulphonate,acmc-209oql,n,n,n-triethylethanaminium-4-methylbenzolsulfonat PubChem CID: 101846 IUPAC Name: 4-methylbenzenesulfonate;tetraethylazanium SMILES: CC[N+](CC)(CC)CC.CC1=CC=C(C=C1)S(=O)(=O)[O-]