missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylammonium p-Toluenesulfonate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 114°C |
| Color | White |
| Quantity | 500 g |
| Formula Weight | 301.45 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tetraethylammonium p-Toluenesulfonate |
Safety and Handling
| EINECSNumber | (2)-0186 |
| TSCA | Yes |