missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylammonium Nitrate 98.0+%, TCI America™
Supplier: TCI America T30825G
Specifications
| Tetraethylammonium Nitrate | |
| 282°C | |
| C8H20N2O3 | |
| 5 g | |
| tetraethylammonium nitrate, ethanaminium, n,n,n-triethyl-, nitrate, ammonium, tetraethyl-, nitrate, ethanaminium, n,n,n-triethyl-, nitrate 1:1, tetraethylazanium nitrate, tetraethyl ammonium nitrate, tetraethylammonium nitrate w/w aqueous solution | |
| [O-][N+]([O-])=O.CC[N+](CC)(CC)CC | |
| 192.26 | |
| 192.26 | |
| Crystalline Powder |
| 1941-26-0 | |
| White | |
| MFCD00041978 | |
| 1479 | |
| JTJKNAJRGLQKDZ-UHFFFAOYSA-N | |
| tetraethylazanium nitrate | |
| 74744 | |
| ≥98.0% (T) |
Chemical Identifiers
| 1941-26-0 | |
| 192.26 | |
| JTJKNAJRGLQKDZ-UHFFFAOYSA-N | |
| 74744 | |
| [O-][N+]([O-])=O.CC[N+](CC)(CC)CC |
| C8H20N2O3 | |
| MFCD00041978 | |
| tetraethylammonium nitrate, ethanaminium, n,n,n-triethyl-, nitrate, ammonium, tetraethyl-, nitrate, ethanaminium, n,n,n-triethyl-, nitrate 1:1, tetraethylazanium nitrate, tetraethyl ammonium nitrate, tetraethylammonium nitrate w/w aqueous solution | |
| tetraethylazanium nitrate |
Safety and Handling
TSCA : Yes