missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Nitrate 98.0+%, TCI America™

Catalog No. p-7137565
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30825G 5 g
T308225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30825G Supplier TCI America Supplier No. T30825G
Only null left

Chemical Identifiers

CAS 1941-26-0
Molecular Formula C8H20N2O3
Molecular Weight (g/mol) 192.26
MDL Number MFCD00041978
InChI Key JTJKNAJRGLQKDZ-UHFFFAOYSA-N
Synonym tetraethylammonium nitrate, ethanaminium, n,n,n-triethyl-, nitrate, ammonium, tetraethyl-, nitrate, ethanaminium, n,n,n-triethyl-, nitrate 1:1, tetraethylazanium nitrate, tetraethyl ammonium nitrate, tetraethylammonium nitrate w/w aqueous solution
PubChem CID 74744
IUPAC Name tetraethylazanium nitrate
SMILES [O-][N+]([O-])=O.CC[N+](CC)(CC)CC

Specifications

Melting Point 282°C
Color White
Quantity 5 g
UN Number 1479
Formula Weight 192.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylammonium Nitrate